Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calretinin Mouse anti-Human, Mouse, Porcine, Rat, Biotin, Clone: 2D7A9, Proteintech
SDP

Catalog No. 501737231 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7231 Supplier Proteintech Group Inc Supplier No. BIOTIN66496100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Calretinin is encoded by the CALB2 gene located on chromosome 16. It is a troponin C superfamily protein having six EF-hand structural motifs. The EF motif allows the protein to bind calcium, and function in calcium signaling, calcium buffering, and modulating neuronal excitability. Calretinin is expressed in the mesothelium, mast cells, some neural cells, hair follicles, and fat cells.

Specifications

Antigen Calretinin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2D7A9
Concentration 0.67 mg/mL
Conjugate Biotin
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CALB2
Gene Accession No. P22676, P47728, Q08331
Gene Alias 29 kDa calbindin, CAB29, CAL2, CALB2, calbindin 2, Calretinin, CR
Gene Symbols Calb2
Host Species Mouse
Immunogen Calretinin Fusion Protein Ag2924
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100127479, 117059, 12308, 794
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.