Learn More
Invitrogen™ Calretinin Polyclonal Antibody
Description
Calretinin Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IHC (PFA)
Specifications
Specifications
| Antigen | Calretinin |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CALB2 |
| Gene Accession No. | P22676, P47728, Q08331, Q3ZBY3 |
| Gene Alias | 29 kDa calbindin; 29kD calretinin; 29kDa calbindin; CAB29; CAL2; Calb2; calbindin 2; calbindin 2, (29kD, calretinin); calbindin 2, 29kDa; calbindin D29K; calretinin; CR; testicular secretory protein Li 8 |
| Gene Symbols | CALB2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.