Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calretinin Polyclonal Antibody (DC8)
GREENER_CHOICE

Catalog No. 180211
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-021-1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-021-1 Supplier Invitrogen™ Supplier No. 180211
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Calretinin, a calcium-binding protein with a molecular weight of 29 kD, is a member of the large family of EF-hand proteins that also includes S-100 protein. This antibody is useful for differentiating mesotheliomas from adenocarcinomas. Positive tissue: Human normal pleura lining/ malignant mesothelioma. Expected Staining Pattern: Cytoplasmic and nuclear.

Calretinin is encoded by the CALB2 gene located on chromosome 16. It is a troponin C superfamily protein having six EF-hand structural motifs. The EF motif allows the protein to bind calcium, and function in calcium signaling, calcium buffering, and modulating neuronal excitability. Calretinin is expressed in the mesothelium, mast cells, some neural cells, hair follicles, and fat cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calretinin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Clone DC8
Conjugate Unconjugated
Dilution Concentrate Antibody
Formulation PBS with 1% BSA and 0.1% sodium azide
Gene CALB2
Gene Accession No. P22676
Gene Alias 29 kDa calbindin; 29kD calretinin; 29kDa calbindin; CAB29; CAL2; Calb2; calbindin 2; calbindin 2, (29kD, calretinin); calbindin 2, 29kDa; calbindin D29K; calretinin; CR; testicular secretory protein Li 8
Gene Symbols CALB2
Host Species Rabbit
Immunogen Recombinant human calretinin.
Positive Control Human normal pleura lining/ malignant mesothelioma
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Clinical
Primary or Secondary Primary
Gene ID (Entrez) 794
Packaging 1 Each
Test Specificity Calretinin
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Host abrv Rb
Reactivity Human
Shipping Condition Wet Ice
Reactivity abrv Hu
Target Gene Calretinin
Validated Application abrv IHC (FFPE)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.