Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calsequestrin Recombinant Rabbit Monoclonal Antibody (JB57-37)
GREENER_CHOICE

Catalog No. PIMA534679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534679 Supplier Invitrogen™ Supplier No. MA534679
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Rat skeletal muscle tissue, mouse skeletal muscle tissue, human fetal skeletal muscle tissue, C2C12.

The sarcoplasmic reticulum (SR) is, in part, responsible for maintaining the level of intracellular calcium in cardiac and skeletal muscle by storing and releasing calcium. Several intralumenal SR calcium binding proteins have been identified, the most prominent of these is calsequestrin. Calsequstrin is a calcium binding protein known to sequester calcium accumulated in the sarcoplasmic reticulum of muscle cells during relaxation and is found discretely localized to the junctional and corbular (terminal cisternae) SR. Calsequestrin functions to localize calcium near the junctional face of the terminal cisternae from which calcium can be released into the cytosol via the ryanodine receptor. This protein is highly acidic and has a large capacity and moderate to low affinity for calcium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calsequestrin
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JB57-37
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CASQ1
Gene Accession No. P31415
Gene Alias Aspartactin; calmitin; calmitine; calsequestrin; calsequestrin 1; calsequestrin 1 (fast-twitch, skeletal muscle); calsequestrin, skeletal muscle isoform; Calsequestrin-1; calsquestrin 1; CASQ; Casq1; CSQ; CSQ1; CSQ-1; Laminin-binding protein; PDIB1; sCSQ; skeletal muscle calsequestrin 1; VMCQA
Gene Symbols CASQ1
Host Species Rabbit
Immunogen Recombinant protein within Human Calsequestrin 1 aa 80-220.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 844
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.