Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calyculin A, >98%, MP Biomedicals™

Catalog No. ICN15837850 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15837850 50 μg
ICN15837810 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15837850 Supplier MP Biomedicals, Inc Supplier No. 0215837850
Add to Cart
Add to Cart

Chemical Identifiers

CAS 101932-71-2
Molecular Formula C50H81N4O15P
Molecular Weight (g/mol) 1009.185
InChI Key FKAWLXNLHHIHLA-OROLNYDDSA-N
Synonym calyculin a
PubChem CID 131632568
IUPAC Name [(2R,3R,5R,7R,8R,9S)-2-[(1S,3S,4S,5R,6R)-14-cyano-3,5-dihydroxy-1-methoxy-4,6,8,9,13-pentamethyltetradeca-7,9,11,13-tetraenyl]-9-[3-[2-[(2S)-4-[[(2S,3S,4S)-4-(dimethylamino)-2,3-dihydroxy-5-methoxypentanoyl]amino]butan-2-yl]-1,3-oxazol-4-yl]prop-2-enyl]-7
SMILES CC1C(CC2(C(C(C(O2)C(CC(C(C)C(C(C)C=C(C)C(=CC=CC(=CC#N)C)C)O)O)OC)OP(=O)(O)O)(C)C)OC1CC=CC3=COC(=N3)C(C)CCNC(=O)C(C(C(COC)N(C)C)O)O)O

Specifications

Assay Percent Range >98%
Quantity 50 μg
Formula Weight 1009.18
Physical Form Lyophilized Powder
Chemical Name or Material Calyculin A
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.