Learn More
CELL MARQUE CORPORATION CAM 5.2 7ML
SDP
Supplier:  CELL MARQUE CORPORATION 452M98
Encompass
Anti-CAM 5.2 is a mouse monoclonal antibody that was generated by using the human colorectal carcinoma cell line HT24. Anti-CAM 5.2 is not technically a pan-keratin antibody because its reactivity has been reported to be restricted primarily to a few keratins. However, since the few keratins are widely expressed in formalin-fixed, paraffin-embedded tissues, especially in simple and glandular epithelia, this antibody can be successfully used as a reliable marker for neoplasms of epithelial differentiation. Anti-CAM 5.2 reacts with the majority of epithelial tumors, including lung, liver, pancreas, GI tract, breast, genitourinary system, female reproductive organs and some endocrine organs.
Catalog No. NC1997089
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.