Learn More
Cambridge Diagnostic Products, Inc.™ Camco™ Stain Pak
Description
- Provides rapid, consistent differential staining of blood smears approximating results obtained from Wright-Giemsa staining
- In addition to highly purified dyestuff and buffers, CAMCO Stain Pak contains important electrolytes that enhance the staining of the nuclei and improve contrast
- Fixative solution is used to stabilize the cell and its contents
- Convenient sizes available
Order Info
Stains are packaged in one-gallon bottles except for Stain Pak, which is packaged in one 473mL bottle each of Fixative, Solution I, and Solution II
Specifications
Specifications
| Quantity | 1 gal. |
| Boiling Point | 100°C |
| Format | Liquid |
| For Use With (Application) | For rapid differential staining of Blood smears |
| Color | Dark Blue |
| Product Type | Solution II (Blue) |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.