Learn More
StatLab Camel Hair Brushes
SDP
Catalog No. 21033413
Catalog No. 21-033-413 Supplier StatLab Supplier No. LWM00903P
Essential in histology analysis, particularly for specimen manipulation and cleaning
  • Uses Include:
  • Dusting microtomes and cryostats: To remove dust and debris from sensitive instruments and samples
  • Catching cryostat sections: To prevent sections from curling and to lay out tissue in water baths
  • Cleaning embedding centers: To ensure clean and accurate embedding of specimens
  • Balancing and cleaning sensitive instruments: To maintain the integrity of instruments used in histology
  • 3. 6.5 in., 12 brushes per pack
Specifications
Product Type Histology Brush
Bristle Material Camel Hair
Bristle Type Stiff
Color-coded No
Handle Material Lacquered
Handle Type Flat
Tip Type Flat
Sterility Non-sterile