Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCU-SCOPE™ Camera and Video Adapters
SDP

The Microscope 0.5x C-Mount Adapter Direct Fit for Unitron's Z730

Supplier:  ACCU-SCOPE™ 4002000E

 View more versions of this product

Catalog No. 01-000-952


Only null left
Add to Cart

Description

Description

  • 0.5x c-mount adapter
  • Direct fit for Z730 only
  • Shipping size 9 x 7 x 5 in.
  • Shipping weight 1lb.
Specifications

Specifications

Camera Adapter
Black
Non-sterile
Not Autoclavable
1.5X
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.