Learn More
Invitrogen™ CaMKII beta Polyclonal Antibody

Description
CaMKII beta Polyclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | CaMKII beta |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 50% glycerol and 0.025% sodium azide |
| Gene | CAMK2B |
| Gene Accession No. | P08413, P28652, Q13554 |
| Gene Alias | Ca++/calmodulin-dependent protein kinase II beta subunit; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II subunit beta; CaM kinase II beta subunit; caM kinase II subunit beta; CAM2; CaMK II; CAMK2; CAMK2B; Camk2d; CAMKB; caMK-II subunit beta; caM-kinase II beta chain; Ck2b; hypothetical protein LOC525416; MGC29528; proline rich calmodulin-dependent protein kinase; β CaMKII; βCaMKII |
| Gene Symbols | CAMK2B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.