Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CaMKII delta Monoclonal Antibody (GT4811)
GREENER_CHOICE

Catalog No. PIMA527735 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527735 Supplier Invitrogen™ Supplier No. MA527735
  • $639.00
    $506.65 / Each of 1

    Save $132.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Positive Control: U87-MG, SK-N-SH, Neuro 2A, C8D30, NIH-3T3, mouse brain, rat brain, transfected 293T Predicted Reactivity: Xenopus laevis (94%), Pig (100%), Rabbit (99%), Chicken (98%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CaMKII delta belongs to the serine/threonine protein kinase family and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells, the enzyme is composed of four different chains: alpha, beta, gamma, and delta. is a delta chain. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Distinct isoforms of this chain have different expression patterns.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaMKII delta
Applications Western Blot
Classification Monoclonal
Clone GT4811
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Camk2d
Gene Accession No. P15791, Q13557, Q6PHZ2
Gene Alias [d]-CaMKII; 2810011D23Rik; 8030469K03Rik; Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II delta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; calcium/calmodulin dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase II, delta; calcium/calmodulin-dependent protein kinase type II delta chain; calcium/calmodulin-dependent protein kinase type II subunit delta; calmodulin-dependent protein kinase II-delta dash; CaM kinase II delta subunit; caM kinase II subunit delta; CAM2; CaMK II; CAMK1; Camk2; CAMK2B; Camk2d; CAMK2G; CAMKB; CAMKD; CAMKG; Camki; CaMK-II delta subunit; CaMK-II subunit delta; CaM-kinase II delta chain; Kiaa4163; RATCAMKI
Gene Symbols Camk2d
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human CaMKII. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108058, 24246, 817
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.