Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CaMKII delta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114939 Supplier Invitrogen™ Supplier No. PA5114939
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CAMK2D.

CaMKII delta belongs to the serine/threonine protein kinase family and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells, the enzyme is composed of four different chains: alpha, beta, gamma, and delta. is a delta chain. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Distinct isoforms of this chain have different expression patterns.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaMKII delta
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Camk2d
Gene Accession No. P15791, Q13557, Q6PHZ2
Gene Alias [d]-CaMKII; 2810011D23Rik; 8030469K03Rik; Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II delta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; calcium/calmodulin dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase II, delta; calcium/calmodulin-dependent protein kinase type II delta chain; calcium/calmodulin-dependent protein kinase type II subunit delta; calmodulin-dependent protein kinase II-delta dash; CaM kinase II delta subunit; caM kinase II subunit delta; CAM2; CaMK II; CAMK1; Camk2; CAMK2B; Camk2d; CAMK2G; CAMKB; CAMKD; CAMKG; Camki; CaMK-II delta subunit; CaMK-II subunit delta; CaM-kinase II delta chain; Kiaa4163; RATCAMKI
Gene Symbols Camk2d
Host Species Rabbit
Immunogen A synthesized peptide derived from human CAMK2D(Accession Q13557), corresponding to amino acid residues R458-I499.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108058, 24246, 817
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.