Learn More
CaMKII delta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
This antibody is specific to the C-term of delta chain, CaMKII delta. It can recognize most isoforms the gene encodes, from 50-72 kDa. It can recognize isoforms 2, 3, 4, 9, 11.
Specifications
Specifications
| Antigen | CaMKII delta |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.19 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CAMK2D |
| Gene Accession No. | Q13557 |
| Gene Alias | CaM kinase II subunit delta, CaMK II subunit delta, CAMK2D, CAMKD |
| Gene Symbols | CAMK2D |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.