Learn More
Selleck Chemical LLC Camostat Mesilate 50mg 59721-29-8 FOY-305
SDP
Supplier:  Selleck Chemical LLC S287450MG
Encompass_Preferred
Camostat (FOY-305) is a trypsin-like protease inhibitor, inhibits airway epithelial sodium channel (ENaC) function with IC50 of 50 nM, less potent to trpsin, prostasin and matriptase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3119
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.