Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Camoteskimab Humanized Recombinant Human Monoclonal Antibody

Catalog No. MA554854
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554854 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA554854 Supplier Invitrogen™ Supplier No. MA554854
Only null left
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

Compatible with Direct ELISA.

Camoteskimab, also known as AVTX-007, is a monoclonal antibody designed to target and inhibit interleukin-18 (IL-18), a cytokine involved in inflammatory processes. By binding to IL-18, camoteskimab prevents its interaction with its receptor, thereby reducing IL-18-mediated signaling pathways that contribute to inflammation. This mechanism of action positions camoteskimab as a potential therapeutic agent for various inflammatory diseases, including atopic dermatitis. Its development is focused on mitigating the effects of excessive IL-18 activity, which is implicated in the pathology of these conditions. As research and clinical trials progress, camoteskimab holds promise for providing a novel treatment option for patients suffering from chronic inflammatory disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Camoteskimab Humanized
Applications ELISA
Classification Recombinant Monoclonal
Concentration 1.87 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias IGIF; IL18; IL1F4
Host Species Human
Immunogen Recombinant Human IL18/IL1F4.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.