Learn More
Genscript Corporation cAMP-HRP
Supplier:  Genscript Corporation M01059
Encompass_Preferred
cAMP-peroxidase conjugate is a product most suitable for GenScript s THETM Anti cAMP monoclonal antibody (Cat No A01509) in immunoassay development for cAMP detection HRP is conjugated to the C-8 position of cAMP
Catalog No. 50-000-56738
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.