Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cAMP Monoclonal Antibody (4H2B6), THE™, GenScript™

Catalog No. 50273599
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-273-599 100 μg
50-273-601 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-273-599 Supplier Genscript Corporation Supplier No. A01509100
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL. Adenosine 3, 5-cyclic monophosphate (cyclic AMP; cAMP) is one of the most important intracellular secondary messengers for transduction events. cAMP is also involved in regulating neuronal, glandular, cardiovascular, immune, and other functions and actions. A number of hormones, such as ACTH, TSH, FSH, and LH, are known to activate cAMP through the action of the enzyme adenylate cyclase, which converts ATP to cAMP. There remains considerable interest in the measurement of intracellular cAMP in tissues and cell cultures, and this may help to provide an understanding of the physiology and pathology of many disease states. Due to the involvement of cAMP in amplifying the response of ligand binding, the second messenger cAMP has been largely employed to monitor the activation of GPCR to facilitate therapeutic drug discovery.GenScript THE™ cAMP Antibody, mAb, Mouse is developed using 3, 5-cyclic AMP-8-KLH as immunogen.

cAMP Monoclonal antibody specifically detects cAMP in Chemical samples. It is validated for ELISA

Specifications

Antigen cAMP
Applications ELISA
Classification Monoclonal
Clone 4H2B6
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene Alias 18 kDa cationic antimicrobial protein; Antibacterial peptide FALL-39; Antibacterial peptide LL-37; CAMP; CAP18; CAP-18; Cathelicidin antimicrobial peptide; cathelin-like protein; Cathelin-related antimicrobial peptide; CLP; Cnlp; Cramp; Cyclic AMP; FALL39; FALL-39; FALL-39 peptide antibiotic; HSD26; LL37; MCLP
Host Species Mouse
Immunogen 3', 5'-cyclic AMP-8-KLH
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG2a κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.