Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ cAMP Parameter Assay Kit
GSA_VASDP

Catalog No. KGE002B Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 Pack
Sufficient For:
1 x 96-well plate
6 x 96-well plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
KGE002B 1 Kit 1 x 96-well plate
SKGE002B 1 Pack 6 x 96-well plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. KGE002B Supplier R&D Systems™ Supplier No. KGE002B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

cAMP Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The cAMP Parameter Assay Kit quantifies multi-species cAMP.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 3.75 to 240 pmol/mL
Assay Sensitivity 8.57 pmol/mL
Conjugate Biotin
Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Cyclic AMP Levels in Various Samples
Sufficient For 1 x 96-well plate
Target Species Multi-species
For Use With (Application) ELISA
Quantity 1 Kit
Storage Requirements Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.