Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ cAMP Parameter Assay Kit
SDP

Catalog No. SKGE002B Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
1 Pack
Sufficient For:
1 x 96-well plate
6 x 96-well plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
SKGE002B 1 Pack 6 x 96-well plates
KGE002B 1 Kit 1 x 96-well plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SKGE002B Supplier R&D Systems™ Supplier No. SKGE002B

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

cAMP Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The cAMP Parameter Assay Kit quantifies multi-species cAMP.

Specifications

Assay Range 3.75 to 240 pmol/mL
Assay Sensitivity 8.57 pmol/mL
Conjugate Biotin
Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Product Line R&D Systems
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Cyclic AMP Levels in Various Samples
Sufficient For 6 x 96-well plates
Target Species Multi-species
For Use With (Application) ELISA
Quantity 1 Pack
Storage Requirements Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.