Learn More
Bioworld Camptothecin, 500 mg
Supplier:  Bioworld 403001492
Encompass_Preferred
A reversible Mitochondrial Topo I inhibitor shown to exhibit antitumour activityAppearance: SolidBoiling Point : 757.01 °C at 760 mmHgDensity : 1.51 g/cm3Melting Point: 260 °C (dec.)(lit.)RTECS Number: UQ0492000
Catalog No. 50-255-694
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.