Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Camptothecin, 97.48%, MP Biomedicals™

Catalog No. icn15973280 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15973280 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15973280 Supplier MP Biomedicals, Inc Supplier No. 0215973280
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7691-02-3
Molecular Formula C8H19NSi2
Molecular Weight (g/mol) 185.42
InChI Key WYUIWUCVZCRTRH-UHFFFAOYSA-N
Synonym 1,1,3,3-tetramethyl-1,3-divinyldisilazane, 1,3-divinyltetramethyldisilazane, 1,3-divinyl-1,1,3,3-tetramethyldisilazane, silanamine, 1-ethenyl-n-ethenyldimethylsilyl-1,1-dimethyl, bis dimethyl vinyl silyl amine, ccris 2450, bis ethenyldimethylsilyl amine, ethenyl-dimethylsilyl amino-dimethylsilyl ethene, n-dimethylvinylsilyl-1,1-dimethyl-1-vinylsilylamine, dvtmds
PubChem CID 82126
IUPAC Name [[[ethenyl(dimethyl)silyl]amino]-dimethylsilyl]ethene
SMILES C[Si](C)(C=C)N[Si](C)(C)C=C

Specifications

Color Yellow
Assay Percent Range 0.9748
Quantity 100 mg
Formula Weight 348.34
Physical Form Solid
Chemical Name or Material Camptothecin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.