Learn More
Description
Biological Activity :Cytotoxic plant alkaloid with antitumor properties; prototypic DNA topoisomerase I inhibitor. Induces single strand DNA breaks and protein-DNA crosslinks
Storage: Store at 4°C.
Note: Inhibitors are guranteed for 1 year from date of receipt.
Specifications
Specifications
| Color | Beige |
| Product Type | Inhibitor |
| Target Proteins | Camptothecin |
| CAS | 3-4-7689 |
| Molecular Formula | C20H16N2O4 |
| Formula Weight | 348.36g/mol |
| Solubility | Soluble to 5mM in DMSO |
| Content And Storage | Store at 4°C |
| Product Line | Novus |
| For Use With (Application) | Induces DNA breaks |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
