Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Campylobacter jejuni Monoclonal Antibody (57-24)
GREENER_CHOICE

Catalog No. 01676877 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-877 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-877 Supplier Invitrogen™ Supplier No. MA135036
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody reacts reacts with a cell wall component of C. jejuni and C. coli. The Hippurate reaction was considered as a marker for C. jejuni. The antibody does not react with C. fetus or C. laridus.

Campylobacter jejuni is a species of curved, long rod-shaped, Gram-negative microaerophilic, bacteria commonly found in animal feces. It is one of the most common causes of human gastroenteritis in the world. C. jejuni is commonly associated with poultry and naturally colonizes the G1 tract of many bird species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Campylobacter jejuni
Applications ELISA
Classification Monoclonal
Clone 57-24
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation Tissue culture supernatant, PBS with 0.1% BSA and 0.02% sodium azide
Gene Alias C. jejuni
Host Species Mouse
Immunogen Proprietary Immunogen.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.