Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAMSAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554999
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554999 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554999 Supplier Invitrogen™ Supplier No. PA554999
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TPTDPGLDSA LEPSGDPHGK CLFDSYRLHD ESNQRTLTLS SSKDANILSE QMSLKEVLDA SVKEVGSSSS DVSGKESVPV EEPLRSRASL IEVDLSDLK Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 65%.

Key microtubule-organizing protein that specifically binds the minus-end of non-centrosomal microtubules and regulates their dynamics and organization (PubMed:19508979, PubMed:21834987, PubMed:24486153, PubMed:24706919, PubMed:24117850). Specifically recognizes growing microtubule minus-ends and stabilizes microtubules (PubMed:24486153, PubMed:24706919). Acts on free microtubule minus-ends that are not capped by microtubule-nucleating proteins or other factors and protects microtubule minus-ends from depolymerization (PubMed:24486153, PubMed:24706919). In contrast to CAMSAP2 and CAMSAP3, tracks along the growing tips of minus-end microtubules without significantly affecting the polymerization rate: binds at the very tip of the microtubules minus-end and acts as a minus-end tracking protein (-TIP) that dissociates from microtubules after allowing tubulin incorporation (PubMed:24486153, PubMed:24706919). Through interaction with spectrin may regulate neurite outgrowth (PubMed:24117850). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAMSAP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAMSAP1
Gene Accession No. Q5T5Y3
Gene Alias 9530003A05Rik; C77823; calmodulin regulated spectrin associated protein 1; calmodulin regulated spectrin-associated protein 1; calmodulin-regulated spectrin-associated protein 1; Camsap1; DKFZp434F195; FLJ31228; PRO2405; RGD1565022
Gene Symbols CAMSAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CAMSAP1. Recombinant protein control fragment (Product #RP-92950).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157922
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.