Learn More
Enzo Life Sciences Canavanine . sulfate (100mg). CAS: 2219-31-0
SDP
Supplier:  Enzo Life Sciences ALX350002M100
Encompass_Preferred
The non-protein amino acid L-canavanine is an analog of L-arginine. Selective inducible nitric oxide synthase (iNOS; NOS II) inhibitor. Induces apoptotic cell death and shows antiproliferative and immunotoxic effects. Alternative name: L-alpha-Amino-gamma-(guanidinooxy)-n-butyric acid . sulfate. Purity: ≥99% (TLC). Appearance: White to-off white powder. Solubility: Soluble in water (100mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8677
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.