Learn More
Cancer Diagnostics, Inc.™ bevel HD Low Profile Coated Microtome Blades
SDP
Catalog No. 22500060 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Quantity:
50 Box
Catalog No. Quantity
22-500-060 50 Box
1 options
This item is not returnable. View return policy
Catalog No. 22-500-060 Supplier Cancer Diagnostics, Inc.™ Supplier No. LPJ3580
Add to Cart
Add to Cart
This item is not returnable. View return policy
outperforms your current low profile blade, every time, every cut
  • The finest steel is paired with proprietary Japanese sharpening and polishing and finished with a specialized PTFE coating to guarantee the quality of every blade for 100% Satisfaction
  • Samples are available
  • Take the Bevel™ Challenge
  • See how this blade outperforms your current low profile blade. Every Time, every cut
  • bevel HD Series
  • Low Profile coated microtome blades
  • Dispenser packs of 50
  • Case of 500 sold in master pack of 10 boxes of 50
Specifications
Profile Low
Material Stainless Steel
Quantity 50 Box
Coating PTFE