Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cancer Diagnostics, Inc.™ Cleanerol™
SDP

Catalog No. 22500214 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Product Type:
Cleanerol Spray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-500-214 Cleanerol Spray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-500-214 Supplier Cancer Diagnostics, Inc.™ Supplier No. CL2525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Base mold release agent

  • Stain and tissue dye remover
  • Paraffin repellent for microtomes, countertops and more
  • 4 oz. (0.2L) bottle

Specifications

Product Type Cleanerol Spray
Color Pale Yellow
For Use With (Application) Stain and tissue dye remover, paraffin cleaner and also used for cutting boards and countertop
Quantity 4 oz. (4/Case)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.