Learn More
Cancer Diagnostics, Inc.™ Eosin Y Histology Secondary Stain
SDP
Catalog No. 22500063 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Quantity:
3800 mL
Catalog No. Quantity
22-500-063 3800 mL
1 options
Catalog No. 22-500-063 Supplier Cancer Diagnostics, Inc.™ Supplier No. EM00PG
Add to Cart
Add to Cart
Used in histology for routine hematoxylin and eosin staining
  • Alcohol based, ready to use secondary stain
  • Filtered prior to bottling
  • Formulated for consistency
  • Eosin Y with Phloxine B produces brighter counter stain
Used in histology for routine hematoxylin and eosin staining
Specifications
For Use With (Application) Used in histology for routine hematoxylin and eosin staining
Quantity 3800 mL
Product Type Eosin Phloxine Stain