Learn More
Cancer Diagnostics, Inc.™ Hematoxylin Staining Reagents
SDP
Catalog No. 22500055 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Product Type:
Gill 1 Hematoxylin
Gill 2 Hematoxylin
Gill 3 Hematoxylin
Harris Hematoxylin
Mayers Hematoxylin Stain
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-500-055 Mayers Hematoxylin Stain
22-500-051 Harris Hematoxylin
22-500-053 Gill 2 Hematoxylin
22-500-052 Gill 1 Hematoxylin
22-500-054 Gill 3 Hematoxylin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22-500-055 Supplier Cancer Diagnostics, Inc.™ Supplier No. CM3951
Add to Cart
Add to Cart
Used in progressive method for histology and cytology preparations
All stains are filtered prior to bottling and use a special process to ensure optimal freshness.
Mercury Free Harris Hematoxylin produces a deep, dark blue-purple stain. It is generally used in regressive methods, but may also be used progressively both in histology and cytology preparations.
Gill 1,2,3 generally used in the progressive method.
Mayer's Hematoxylin used for routine H&E and immunochemistry.
Used in progressive method for histology and cytology preparations
Specifications
For Use With (Application) Used for routine H and E and immunochemistry
Quantity 3800mL
Product Type Mayers Hematoxylin Stain