Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cancer Diagnostics, Inc.™ Ink Aid
SDP

Catalog No. 22500211 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Quantity:
5 Case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-500-211 5 Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-500-211 Supplier Cancer Diagnostics, Inc.™ Supplier No. IA4000
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Tissue marking dye setting solution for better adherence.

  • Tissue marking dye setting solution
  • Spray onto dyed tissue for better adherence through solutions

Specifications

Boiling Point 118.1°C
Capacity 8 oz.
Color Clear
Melting Point 16.7°C
Product Type Tissue Marking Dye
Quantity 5 Case
Solution Type Buffer
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.