Learn More
Cancer Diagnostics, Inc.™ Molded 1-Piece Biopsy Cassettes (Square Holes) - Taped
SDP
Catalog No. p-4255253 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lavender
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-500-602 Gray
22-500-601 White
22-500-603 Blue
22-500-604 Green
22-500-605 Pink
22-500-606 Yellow
22-500-608 Lavender
22-500-609 Peach
22-500-610 Aqua
22-500-611 Tan
22-500-607 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 22-500-602 Supplier Cancer Diagnostics, Inc.™ Supplier No. BHTP1001
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Taped cassettes optimized for automated cassette printers
  • Excellent adherence for all inks
  • Optimal polymer technology resistant to all chemicals
  • Easy optimal writing surface
  • Available in 11 colors
Specifications
Pore Style Square
Color Gray
Product Type Biopsy Cassette