Learn More
Cancer Diagnostics, Inc.™ Nylon Mesh Biopsy Bags
SDP
Catalog No. 22500469 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Color:
White
Yellow
Quantity:
Pack of 100
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
22-500-469 White Pack of 100
22-500-471 White Pack of 1000
22-500-470 Yellow Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-500-469 Supplier Cancer Diagnostics, Inc.™ Supplier No. BIO3045W
Add to Cart
Edge
Add to Cart
Reduce cross-contamination
  • Made from nylon mesh with precise mesh diameter
  • Allow tiny specimens to be captured by straining the fixative when poured into the bag
  • Can be used to contain tiny biopsies during decalcification
  • Simply insert specimen, fold bag to enclose cassette or capsule, and process
Order Info
Measure 30 x 45mm; available in white and yellow.
Specifications
Color White
Dimensions (L x W) 30 x 40 mm
Quantity Pack of 100