Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cancer Diagnostics, Inc.™ Pureview Staining Reagents
SDP

Catalog No. 22500059 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Quantity:
500mL
3800mL
Product Type:
Alcoholic Eosin Y Stain
Focus Stain
Hematoxylin 660+ Stain
PH Blue Stain
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-500-059 3800mL PH Blue Stain
22-500-056 500mL Hematoxylin 660+ Stain
22-500-057 500mL Alcoholic Eosin Y Stain
22-500-058 3800mL Focus Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-500-059 Supplier Cancer Diagnostics, Inc.™ Supplier No. 167024
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pureview is a hematoxylin and eosin staining system consisting of Hematoxylin 660+, Eosin 615, PH Blue, and Focus. Each component must pass rigorous analytical standards and are formulated to help standardize the H&E staining process with repeatable, consistent results.

Hematoxylin 660+™ A certified, pre-filtered progressive hematoxylin. Hematoxylin 660+ is formulated to stain more slides than traditional hematoxylins and produces crisp, well delineated nuclear chromatin.
Eosin 615™ A pre-filtered optimal differentiator. Red blood cells stain red, and connective tissue stains varying shades of pink to orange.
PH Blue™ A bluing reagent buffered to a constant pH 8. Fluctuations in pH have been associated with floaters and unclear nuclear detail. pH Blue is designed to be unaffected by carryover and is formulated to help standardize the staining process.
Focus™ A clarifier that removes non-specific staining and background haze. Focus is formulated to sharpen nuclear detail producing a finishing effect that optimizes nuclear detail.

Specifications

For Use With (Application) Used for staining
Quantity 3800mL
Product Type PH Blue Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.