Learn More
Cancer Diagnostics, Inc.™ Vortex™ Biopsy Cassettes
SDP
Catalog No. 22500886 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Packaging Type:
Loose
Taped
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Packaging Type
22-500-886 Blue Loose
22-500-885 White Loose
22-500-887 Green Loose
22-500-890 White Taped
22-500-888 Pink Loose
22-500-891 Blue Taped
22-500-893 Pink Taped
22-500-892 Green Taped
22-500-894 Yellow Taped
22-500-889 Yellow Loose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 22-500-886 Supplier Cancer Diagnostics, Inc.™ Supplier No. VB1002
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for routine processing and embedding of tissues and cells during histological studies
  • Rounded interiors without corners
  • One-piece breakable lid design
  • Filtration enhanced by side vents
  • Processed biopsies are easy to find
  • Small and assorted colors available
  • Loose and taped options for packaging
Specifications
Packaging Type Loose
Color Blue
Product Type Vortex Biopsy Cassette