Learn More
Invitrogen™ Candida albicans Polyclonal Antibody

Description
PA1-27158 detects C. albicans and other yeasts from yeast samples.
Specifications
Specifications
| Antigen | Candida albicans |
| Applications | ELISA, Immunodiffusion, Immunoelectrophoresis, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | C. albicans |
| Host Species | Rabbit |
| Immunogen | Candida albicans, type A. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.