Learn More
ACROBiosystems ACROBiosystems Canine 4-1BB / TNFRSF9 Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems 41BC52531MG
Encompass_Preferred
Recombinant Protein;1MG;Canine 4-1BB Protein, Fc Tag (41B-C5253) is expressed from human 293 cells (HEK293). It contains AA Ile 24 - Ser 185 (Accession # XP_850336.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.4 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 4-1BB Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. 41L-H5254) at 5 μg/mL (100 μL/well) can bind Canine 4-1BB Protein, Fc Tag (Cat. No. 41B-C5253) with a linear range of 0.3-5 ng/mL (QC tested).
Catalog No. 50-210-6424
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.