Learn More
Innovative Research Inc CANINE ALBUMIN PURIFIED LYOPH
Supplier:  Innovative Research Inc ICNALBLY100MG
Encompass_Preferred
Canine Albumin Purified Lyophilized 100mg.Species: Canine. Target: AlbuminPurification Method: Chromatographically purifiedPurity: >95% as determined by SDS-PAGE AnalysisSource: Dog plasma
Catalog No. NC2121594
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.