Learn More
Description
- NGF nerve growth factor [ Canis lupus familiaris (dog) ]
- Also known as: NGFB
- The beta-NGF insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- NGF is a member of the NGF-beta family and is a secreted protein which homodimerizes and is incorporated into a larger complex.
- It has nerve growth stimulating activity and the complex is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons.
- Mutations in NGF have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NFG expression is associated with allergic rhinitis.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | caNGF |
| Accession Number | NP_001181879 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 739 bp |
| Gene ID (Entrez) | 4803 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Canine |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
