Learn More
*HELICA BIOSYSTEMS CANINE CRP QUANTITATIVE
Supplier:  *HELICA BIOSYSTEMS 931CRP01D96
Encompass_Preferred
Clinical Diagnostic Test Kits; C-Reactive Protein Assay, ELISA Test Kit, Canine CRP Quantitative detection, 96-strip microwell plate based assay, Results in 1.5 hours, One-year stability; Excellent reproducibility
Catalog No. 50-980-599
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.