Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Canine Distemper Virus Monoclonal Antibody (I47G)
GREENER_CHOICE

Catalog No. PIMA17468
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17468 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17468 Supplier Invitrogen™ Supplier No. MA17468
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7468 antibody reacts with Canine Distemper virus in viral and infected tissue samples. Non-reactive with: Canine Parvovirus and Canine Coronavirus MA1-7468 has been successfully used in ELISA applications. The MA1-7468 antibody was raised against canine distemper virus viral preparation.

This antibody recognizes canine Distemper Virus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Canine Distemper Virus
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone I47G
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias CDV
Host Species Mouse
Immunogen Viral preparation.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.