Learn More
Kingfisher Biotech Inc Canine Fas Ligand Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1088D005
Encompass
The Canine Fas Ligand yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine Fas Ligand applications are for cell culture, ELISA standard, and Western Blot Control. Canine Fas Ligand yeast-derived recombinant protein can be purchased in multiple sizes. Canine Fas Ligand Specifications: (Molecular Weight: 17.3 kDa) (Amino Acid Sequence: QIGQPNPPSE KRELRKVAHL TGKPNSRSIP LEWEDTYGIA LVSGVKYKKG GLVINDTGLY FVYSKVYFRG QSCNNKPLNH KVYMRNSKYP QDLMLMEGKI MNYCTTGQMW ARSSYLGAVF NLTSADHLYV NVSELSLVSF EESKTFFGLY KL (152)) (Gene ID: 442968). For research use only. Made in the USA
Catalog No. 50-253-3698
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.