Learn More
Invitrogen™ Canine/Feline Parvovirus Monoclonal Antibody (CPVI-2AI)
Shop All Thermo Scientific Products

Description
MA1-82094 detects canine parvovirus (CPV) and feline panleucopenia virus (FPV), and will also cross-react with mink parvovirus. This antibody does not recognize canine adenovirus (type2), canine coronavirus, canine distemper, canine parinfluenza virus, feline leukaemia virus, or feline immunodeficiency virus This product does not require protein digestion pre-treatment of paraffin embedded sections. This product does not require antigen retrieval using heat treatment prior to staining of paraffin embedded sections. Mouse anti Canine/ Feline parvovirus antibody, clone CPV1-2A1 recognizes canine parvovirus (CPV) and feline panleucopenia virus (FPV).
Specifications
Specifications
| Antigen | Canine/Feline Parvovirus |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CPVI-2AI |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with <0.1% sodium azide; pH 7.2 |
| Gene Alias | CPV; FPV; parvo |
| Host Species | Mouse |
| Immunogen | Proprietary Immunogen. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.