Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Canine Heartworm Monoclonal Antibody (H21G)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA17012
Description
MA1-7012 detects Canine Heartworm. MA1-7012 has been successfully used in ELISA, immunofluorescence, and Western blot procedures. The MA1-7012 immunogen is whole Canine Heartworm.
Canine heartworm is a common parasite of dogs worldwide. Heartworm occurs in about 30 other species as well but rarely affects humans. The infection is commonly transmitted by mosquitoes. Adult heartworms can grow to be up to 10 inches in length and can live for 5-7 years in dogs affecting the heart and lungs.
Specifications
Canine Heartworm | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Nematode | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
H21G | |
Unconjugated | |
Dirofilaria; dirofilariasis; Filaria | |
Whole Canine Heartworm lysate. | |
1 mL | |
Primary | |
4°C | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction