Learn More
Kingfisher Biotech Inc Canine IFN gamma Biotinylated Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RPB1863D010
Encompass
The Canine IFN gamma Biotinylated yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine IFN gamma Biotinylated applications are for cell culture. Canine IFN gamma Biotinylated yeast-derived recombinant protein can be purchased in multiple sizes. Canine IFN gamma Biotinylated Specifications: (Molecular Weight: 16.7 kDa) (Amino Acid Sequence: QAMFFKEIEN LKEYFNASNP DVSDGGSLFV DILKKWREES DKTIIQSQIV SFYLKLFDNF KDNQIIQRSM DTIKEDMLGK FLNSSTSKRE DFLKLIQIPV NDLQVQRKAI NELIKVMNDL SPRSNLRKRK RSQNLFRGRR ASK (143)) (Gene ID: 403801). For research use only. Made in the USA
Catalog No. 50-253-2341
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.