Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Canine IL-10 Quantikine ELISA Kit
SDP

Catalog No. CA1000 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
CA1000 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CA1000 Supplier R&D Systems™ Supplier No. CA1000

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Canine IL-10 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Canine IL-10 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies canine IL-10 in cell culture supernates (25 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 3.8pg/mL
Conjugate HRP
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Canine IL-10
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 403628
Sample Volume Cell Culture Supernates (25uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10 mgC126451, interleukin 10, interleukin-10, TGIF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.