Learn More
Description
Includes
Canine IL-17/IL-17A Capture Antibody; Canine IL-17/IL-17A Biotinylated-conjugated Detection Antibody
Contains the basic components required to develop an ELISpot (quantitative sandwich ELISA) assay
Optimized capture and detection antibody pairings and recommended concentrations save lengthy development time
Generic development protocols provide directi
Specifications
Specifications
| Cross Reactivity | Dog |
| Quantity | 1 Kit |
| Synonym | CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Form | Lyophilized |
| Product Line | R&D Systems |
| Product Type | ELISpot Development Module |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
