Learn More
Description
- Interleukin 17A (IL17A), a proinflammatory cytokine secreted by activated T-lymphocytes, is a potent inducer of the maturation of CD34-positive hematopoietic precursors into neutrophils.
- IL17RA is a type I membrane glycoprotein that binds to interleukin 17A with low affinity.
- It has a multimeric structure and plays a pathogenic role in many inflammatory and autoimmune diseases.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | caIL17RA |
| Accession Number | XP_025317395 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2608 bp |
| Gene ID (Entrez) | 23765 |
| Gene Alias | CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17 R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17 RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Canine |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
