Learn More
Specifications
Specifications
| Assay Range | 0.31 - 20 ng/mL |
| Product Line | R&D Systems |
| Product Type | Solid Phase Sandwich ELISA |
| Gene ID (Entrez) | 3606 |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 |
| Target | Canine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
