Learn More
Kingfisher Biotech Inc Canine IL-1 beta ELISA
SDP
Supplier:  Kingfisher Biotech Inc DIY0679D003
Encompass_Preferred
The Canine IL-1 beta ELISA contains capture antibody protein standard and detection antibody for development of a Canine IL-1 beta ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers concentrations incubation times incubation temperatures and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Canine IL-1 beta ELISA can also be used to measure Dingo IL-1 beta. Alternate names include IL1B IL-1 IL1-BETA IL1F2 interleukin 1 beta IL1beta. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5141
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.