Learn More
Description
- An economical alternative to ELISpot Kits
- Optimized capture and detection antibody pairings and recommended concentrations save lengthy development time
- Generic development protocols provide direction to start an optimization protocol
- Customize the assay to your specific needs
Module content:
- Capture Antibody
- Biotinylated-conjugated Detection Antibody
Specifications
Specifications
| Quantity | 1 Kit |
| Synonym | B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
